Learn More
Rockland Immunochemicals TrueBlot Biotin Magnetic Beads 5 mL
SDP
Catalog No. 501943960
Encompass_Preferred
TrueBlot Magnetic Beads are uniform non-aggregating super-paramagnetic beads consisting of a ferric oxide core functionalized with various silane groups. The super-paramagnetic nanoparticles are coupled with a biomolecule such as Biotin and are specifically designed tested and quality controlled for magnetic separation isolation and purification of avidin and streptavidin-labeled components using manual or automatic platforms. Biotin magnetic beads are stable pre-blocked beads with high binding capacity that provide rapid and efficient biomolecule purification from complex samples. Bead mean diameter is ~0.5 μm bead concentration is 5 mg/mL and binding capacity is ≥ 30 µg streptavidin/mg of beads.
Catalog No. 50-194-3960 Supplier Rockland Immunochemicals Supplier No. B000185
TrueBlot Magnetic Beads are uniform non-aggregating super-paramagnetic beads consisting of a ferric oxide core functionalized with various silane groups. The super-paramagnetic nanoparticles are coupled with a biomolecule such as Biotin and are specifically designed tested and quality controlled for magnetic separation isolation and purification of avidin and streptavidin-labeled components using manual or automatic platforms. Biotin magnetic beads are stable pre-blocked beads with high binding capacity that provide rapid and efficient biomolecule purification from complex samples. Bead mean diameter is ~0.5 μm bead concentration is 5 mg/mL and binding capacity is ≥ 30 µg streptavidin/mg of beads.