Learn More
Seracare Life Sciences Inc TRUEBLUE HRP SUBSTRATE
Catalog No. 5067581
Encompass_Preferred
TrueBlue is up to 100 times more sensitive than DAB and it is free of known carcinogens. The strong blue reaction product provides excellent contrast with DAB and other substrates. HistoMark TrueBlue provides the most sensitive means available to detect peroxidase-labeled conjugates. Because of its high sensitivity, TrueBlue is recommended for detection of the least abundant marker in immunohistochemistry applications. TrueBlue is alcohol and xylene insoluble.
This item is not returnable. View return policy
Catalog No. 50-675-81 Supplier Seracare Life Sciences Inc Supplier No. 55100049
TrueBlue is up to 100 times more sensitive than DAB and it is free of known carcinogens. The strong blue reaction product provides excellent contrast with DAB and other substrates. HistoMark TrueBlue provides the most sensitive means available to detect peroxidase-labeled conjugates. Because of its high sensitivity, TrueBlue is recommended for detection of the least abundant marker in immunohistochemistry applications. TrueBlue is alcohol and xylene insoluble.