Learn More
Seracare Life Sciences Inc TRUEBLUE HRP SUBSTRATE KIT
Catalog No. 5067518
Encompass_Preferred
This kit provides the most sensitive detection for peroxidase systems utilizing KPL's proprietary TrueBlue Substrate. In our studies, TrueBlue is 50-100 times more sensitive than DAB and is non-carcinogenic. Strongly recommended for the detection of least-abundant markers. TrueBlue forms a blue chromogenic product, providing excellent contrast with DAB and other substrates for multiple labeling experiments.
This item is not returnable. View return policy
Catalog No. 50-675-18 Supplier Seracare Life Sciences Inc Supplier No. 55100035
This kit provides the most sensitive detection for peroxidase systems utilizing KPL's proprietary TrueBlue Substrate. In our studies, TrueBlue is 50-100 times more sensitive than DAB and is non-carcinogenic. Strongly recommended for the detection of least-abundant markers. TrueBlue forms a blue chromogenic product, providing excellent contrast with DAB and other substrates for multiple labeling experiments.