Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Drucker Diagnostics Truebond Veterinary Centrifuge

Catalog No. 21100510 Shop All Drucker Diagnostics Products
Change view
Click to view available options
Max. RCF:
1,850 xg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Max. RCF
21-100-510 1,850 xg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-100-510 Supplier Drucker Diagnostics Supplier No. 00376009005
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Veterinary centrifuge for blood, urine, fecal, and microhematocrit.

  • Spin blood, urine, fecal, and crit in one centrifuge
  • Program up to 10 custom settings using the digital display
  • Compact design requires just 1.35 ft² (1,254 cm²) of counter space
  • Capacity: 6 blood, urine, or fecal tubes. 8 microhematocrit tubes.
  • Max RCF: 1,850 xg
  • Max Speed: 3,900 RPM
  • Applications: Microhematocrit, Serum, Urine, Fecals 

Specifications

Capacity 6x blood tubes (.5-15 mL), 8x microhematocrits
Max. RCF 1,850 xg
Max. Speed 3,900 Max RPM
Includes Rotor (Fixed Angle), 6x .5-10mL Tube Holders, 6x 15mL Tube Holders, 2x Microhematocrit Carriers
Noise Level 64 dBA
Rotor Included Yes
Net Weight 12 lbs (5.4 kg)
Electrical Requirements 95 to 253 V 50/60 Hz
Product Type Multipurpose Veterinary Centrifuge
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.