Learn More
Hach Company Truecal - Calibration Software
Catalog No. 5000207078 Shop All Hach Company Products
Encompass_Preferred
Truecal software to upgrade DR3900 or DR6000 spectrophotometers older than March 2018 Truecal from Hach reduces variations in results caused by variances in chemistry raw materials A bar code contains the calibration curve data specific to each batch of chemistry and automatically updates the calibration curve With TNTplus products using the Truecal feature you ll have one less thing to worry about during crucial proficiency testing or permit-limit compliance testing
Catalog No. 50-002-07078 Supplier Hach Company Supplier No. LPZ440.99.00001
Truecal software to upgrade DR3900 or DR6000 spectrophotometers older than March 2018 Truecal from Hach reduces variations in results caused by variances in chemistry raw materials A bar code contains the calibration curve data specific to each batch of chemistry and automatically updates the calibration curve With TNTplus products using the Truecal feature you ll have one less thing to worry about during crucial proficiency testing or permit-limit compliance testing