Learn More
SW™ Sustainability Solutions TrueForm™ TF-95BK Black 3.5 mil Nitrile Exam Gloves - 200 ct
SDP
Catalog No. 18001048 Shop All SW Sustainability Solutions Products
Catalog No. 18-001-048 Supplier SW™ Sustainability Solutions Supplier No. N191886
Only null left
Add to Cart
Add to Cart
Designed for food safety, these gloves combines sustainability with exceptional barrier protection delivering superior grip, dexterity, and comfort for clean, safe handling in demanding tasks.
  • EcoTek™ Sustainable Technology
  • Finger textured surface for excellent wet and dry grip
  • Tear-resistant beaded cuff eases donning
  • Silicone-Free eliminates risk of contamination in sensitive food processes
  • Exceeds 1.5 AQL Standards-Provides consistent reliability and reduced risk of defects
  • Comfortable Ergonomic Fit-Designed for all-day wear with excellent dexterity and tactile sensitivity
  • The right weight for resiliency and dexterity
  • SHA Dermatologically Approved
TRUSTED_SUSTAINABILITY
EcoTek gloves tested to ASTM D5526-12 Standard Test Method for Determining Anaerobic Biodegradation of Plastic Materials Under Accelerated Landfill Conditions. Results are 92.6% biodegradation in 60% solids landfill at 945 days. Future results cannot be predicted/extrapolated.