Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SW™ Sustainability Solutions TrueForm™ TF-95BL Blue 3.5 mil Sustainable Nitrile Exam Gloves
SDP

Catalog No. 19021876 Shop All SW Sustainability Solutions Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-021-876 X-Small
19-021-877 Small
19-021-878 Medium
19-021-879 Large
19-021-880 X-Large
19-021-881 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-021-876 Supplier SW™ Sustainability Solutions Supplier No. N091341
Add to Cart
Add to Cart

Value and performance hand protection with EcoTek™ sustainable technology.

  • EcoTek™ sustainable technology
  • EnerGel™ Hand Conditioning Technology clinically proven to hydrate and soothe skin
  • Proprietary coating actively manages the skin’s natural pH balance™ to discourage microbial growth that can lead to infection
  • Finger textured surface for wet and dry grip
  • Low Dermatitis Potential (LDP) is an FDA approved claim that gloves were clinically tested and have a reduced potential to cause skin irritation
  • Exceeds 1.5 AQL laboratory standards
TRUSTED_SUSTAINABILITY

Specifications

Accelerator Free No
Applications Healthcare, Laboratory
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Fingertip Textured
Finger Thickness (English) 3.5 mil
Finger Thickness (Metric) 0.08 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 2.4 mil
Palm Thickness (Metric) 0.06 mm
Powdered Powder Free
Product Type Nitrile Examination Glove
Size X-Small
Sterility Non-sterile
Thickness (English) 2.4 mil
Thickness (Metric) 0.06 mm
Model N091341
Series TF-95RB
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.