Learn More
SW™ Sustainability Solutions TrueForm™ TF-95NV Navy 3.5 mil Nitrile Exam Gloves - 200 ct
SDP
Catalog No. 18001037 Shop All SW Sustainability Solutions Products
Change view
Click to view available options
Size:
Small
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-001-037 -
18-001-038 Small
18-001-039 -
18-001-040 -
18-001-041 -
18-001-042 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-001-037 Supplier SW™ Sustainability Solutions Supplier No. N191341
Add to Cart
Add to Cart
Designed for daily tasks, these nitrile exam gloves deliver reliable protection with a sustainable edge.
  • EcoTek™ Sustainable Technology
  • Finger textured surface for excellent wet and dry grip
  • Tear-resistant beaded cuff eases donning
  • SHA Dermatologically Approved
  • Low Dermatitis Potential (LDP) is an FDA approved claim that gloves were clinically tested and have a reduced potential to cause skin irritation
  • Tested for use with chemotherapy drugs
  • Silicone-free eliminates risk of contamination in sensitive food processes
  • Exceeds 1.5 AQL standards providing consistent reliability and reduced risk of defects
  • Comfortable ergonomic fit designed for all-day wear with excellent dexterity and tactile sensitivity
  • The right weight for resiliency and dexterity
TRUSTED_SUSTAINABILITY
EcoTek gloves tested to ASTM D5526-12 Standard Test Method for Determining Anaerobic Biodegradation of Plastic Materials Under Accelerated Landfill Conditions. Results are 92.6% biodegradation in 60% solids landfill at 945 days. Future results cannot be predicted/extrapolated.