Learn More
Sigma Aldrich Fine Chemicals Biosciences TrueGel3D Hydrogel Kits fA
Catalog No. 502905372
Encompass
The gel is formed by stable thioether bonds between maleimide groups of dextran (fAST-DEXTRAN) and thiol groups on CD cell-degradable crosslinker. Malemide-functionalized dextran is specifically used to decrease the time for gel formation. The dextran polymer does not have cell attachment sites but it can be customized to conjugate RGD peptide using TrueGel3D RGD peptide (TRUERGD-1UM) prior to the crosslinking step. The CD cell-degradable crosslinker is composed of matrix metalloprotease (MMP)-cleavable peptide (Pro-Leu-Gly-Leu-Trp-Ala) which allows cells to spread and migrate by secreting matrix metalloproteases (MMP1 MMP3 MMP7 and MMP9).TrueGel3D enzymatic cell recovery SOLUTIONon (TRUEENZ-500UL) is used to dissolve dextran polymers for easy recovery of chemically fixed or live cells for post-culture analyses including RT-PCR western blots and other molecular biological studies.
Catalog No. 50-290-5372 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. TRUE31KT
The gel is formed by stable thioether bonds between maleimide groups of dextran (fAST-DEXTRAN) and thiol groups on CD cell-degradable crosslinker. Malemide-functionalized dextran is specifically used to decrease the time for gel formation. The dextran polymer does not have cell attachment sites but it can be customized to conjugate RGD peptide using TrueGel3D RGD peptide (TRUERGD-1UM) prior to the crosslinking step. The CD cell-degradable crosslinker is composed of matrix metalloprotease (MMP)-cleavable peptide (Pro-Leu-Gly-Leu-Trp-Ala) which allows cells to spread and migrate by secreting matrix metalloproteases (MMP1 MMP3 MMP7 and MMP9).TrueGel3D enzymatic cell recovery SOLUTIONon (TRUEENZ-500UL) is used to dissolve dextran polymers for easy recovery of chemically fixed or live cells for post-culture analyses including RT-PCR western blots and other molecular biological studies.