Learn More
Sigma Aldrich Fine Chemicals Biosciences TrueGel3D Hydrogel Kits SL
Catalog No. 502900664
Encompass
The gel is formed by stable thioether bonds between thiol groups on PEG non cell-degradable crosslinker and thio-reactive groups on SLO-DEXTRAN. The polymer SLO-DEXTRAN is functionalized with thiol-reactive groups which react with crosslinker at slower rate than maleimides in fAST-DEXTRAN. SLO-DEXTRAN does not have sites for cell attachment but it can be customized to conjugate RGD peptide using TrueGel3D RGD peptide (TRUERGD-1EA) prior to the crosslinking step.TrueGel3D enzymatic cell recovery SOLUTIONon (TRUEENZ-500UL) is used to dissolve dextran polymers for easy recovery of chemically fixed or live cells for post-culture analyses including RT-PCR western blots and other molecular biological studies.
Catalog No. 50-290-0664 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. TRUE61KT
The gel is formed by stable thioether bonds between thiol groups on PEG non cell-degradable crosslinker and thio-reactive groups on SLO-DEXTRAN. The polymer SLO-DEXTRAN is functionalized with thiol-reactive groups which react with crosslinker at slower rate than maleimides in fAST-DEXTRAN. SLO-DEXTRAN does not have sites for cell attachment but it can be customized to conjugate RGD peptide using TrueGel3D RGD peptide (TRUERGD-1EA) prior to the crosslinking step.TrueGel3D enzymatic cell recovery SOLUTIONon (TRUEENZ-500UL) is used to dissolve dextran polymers for easy recovery of chemically fixed or live cells for post-culture analyses including RT-PCR western blots and other molecular biological studies.