Learn More
Sigma Aldrich Fine Chemicals Biosciences TrueGel3D RGD Integrin Adh
Catalog No. 502905416
Encompass
RGD sequences on extracellular matrix proteins (fibronectin vitronectin) promote cell adhesion to cellular integrin receptors. The thiol group on the N-terminus of TrueGel3D RGD peptide acts as linker to bind RGD motif to the polymer (Dextran/PVA). The conjugated RGD motif on polymer promotes adhesion of cells with appropriate integrin receptors.
Catalog No. 50-290-5416 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. TRUERGD3EA
RGD sequences on extracellular matrix proteins (fibronectin vitronectin) promote cell adhesion to cellular integrin receptors. The thiol group on the N-terminus of TrueGel3D RGD peptide acts as linker to bind RGD motif to the polymer (Dextran/PVA). The conjugated RGD motif on polymer promotes adhesion of cells with appropriate integrin receptors.