Learn More
Sigma Aldrich Fine Chemicals Biosciences TrueNorthR Cardboard Micro
Supplier:  Sigma Aldrich Fine Chemicals Biosciences HS120220
Encompass
Low profile boxes are available in two sizes to accomodate standard 0.5 and 0.2 mL microtubes and vessels of a similar size. (must use tubes capable of being sotred at low temperatures)
Catalog No. 50-290-1515