Learn More
Sigma Aldrich Fine Chemicals Biosciences TrueNorthR Cool Container
Supplier:  Sigma Aldrich Fine Chemicals Biosciences HS28729R
Encompass
These durable containers for chilling temperature-sensitive samples have heavy insulation properties for the ultra cold. Cool containers are ideal for ice and ice-salt slurries.
Catalog No. 50-290-0852