Learn More
Sigma Aldrich Fine Chemicals Biosciences TrueNorthR Cool Container
Catalog No. 502900435
Encompass
These durable containers for chilling temperature-sensitive samples have heavy insulation properties for the ultra cold. Cool containers are ideal for ice and ice-salt slurries.
Catalog No. 50-290-0435 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. HS28724B
These durable containers for chilling temperature-sensitive samples have heavy insulation properties for the ultra cold. Cool containers are ideal for ice and ice-salt slurries.