Learn More
Sigma Aldrich Fine Chemicals Biosciences TrueNorthR Cryogenic Vial
Supplier:  Sigma Aldrich Fine Chemicals Biosciences HS23202N500EA
Encompass
The True North Cryogenic Vials innovative design saves space in your freezer. The vials are non-pyrogenic non-cytotoxic and DNase RNase free.
Catalog No. 50-290-4876