Learn More
Sigma Aldrich Fine Chemicals Biosciences TrueNorthR Mini Cooler for
Supplier:  Sigma Aldrich Fine Chemicals Biosciences HS120075
Encompass
Protect temperature sensitive samples at the bench or in the freezer. Mini Coolers are ideal for keeping PCR reagents enzymes and other biochemical reagents cool while working at the bench. filled with insulating gel the mini coolers can also be used for ccontrolled sample freezing and for longer temperature retention samples.
Catalog No. 50-290-4531