Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TrueTag™ Donor DNA Kit, HA

Catalog No. A53809
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A53809 10 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A53809 Supplier Invitrogen™ Supplier No. A53809
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

The Invitrogen TrueTag Donor DNA Kit, HA, provides a rapid solution to drive high-efficiency gene tagging (up to 100% edited cells) with transfection-ready donor DNA for genome editing experiments.

  • A rapid solution to drive high-efficiency gene tagging (up to 100% edited cells) with transfection-ready donor DNA for genome editing experiments
  • Kit provides everything needed to produce a high-quality DNA donor to tag the gene with a selection marker and HA
  • Simply provide two locus-specific primers to add homology arms, PCR-amplify, and purify for a transfection-ready donor in hours, not days
  • One-step PCR amplification to generate donor templates – no cloning required
  • Enrich up to 100% HA-tagged cells

Each kit is designed to enable users to:

  • Tag the carboxy- (C-) or amino- (N-) terminus of the gene of interest
  • Choose from puromycin or blasticidin selection markers – whatever best suits the cell model
  • Easily remove antibiotic resistance markers with Cre/Lox following cell enrichment

Included in the kit:

  • Four linear donor templates: N-terminus/puromycin resistance, N-terminus/blasticidin resistance, C-terminus/puromycin resistance, C-terminus/blasticidin resistance
  • Phusion Flash High-Fidelity PCR Master Mix
  • PCR cleanup columns and buffers
  • Positive control primers to tag human beta-actin (ACTB)

Intuitive software to rapidly design gene-specific reagents

Target-specific gene editing reagents are required for proper knock-in of your fluorescent tag. The Invitrogen TrueDesign Genome Editor web-based software does the design work for you:

  1. Identifies target gene transcript
  2. Designates insertion site
  3. Designs and enables easy ordering of all necessary components

Required reagents available via TrueDesign software (not included in kit):

  • CRISPR guide RNA or TALENs designed adjacent to N- or C-terminus
  • Oligos to PCR-amplify the donor and add appropriate homology arms
  • Sequencing primers for assessment of CRISPR-Cas9 cleavage efficiency
  • Verification primers to validate proper insertion of the TrueTag DNA Donor

Also required and available for purchase separately or as part of TrueDesign workflow:

  • TrueCut Cas9 Protein v2, TrueCut HiFi Cas9 Protein, or GeneART CRISPR mRNA
  • CRISPRMAX Transfection Reagent or MessengerMAX Transfection Reagent
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Protein Detection and Isolation
Quantity 10 Reactions

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.