Learn More
Vector Laboratories Vector TrueVIEW Autofluorescence Quenching Kit, 15ml
SDP
Catalog No. NC2023586
Encompass
The TrueVIEW Autofluorescence Quenching Kit provides a novel way to diminish unwanted autofluorescence from non-lipofuscin sources and dramatically improve signal-to-noise ratio. TrueVIEW removes unwanted autofluorescence in tissue sections due to aldehyde fixation, the presence of red blood cells, and structural elements such as collagen and elastin. The quenching action of the kit reagents provides a clear, unambiguous, “true view” localization of the target antigen.
Catalog No. NC2023586 Supplier Vector Laboratories Supplier No. SP840015
The TrueVIEW Autofluorescence Quenching Kit provides a novel way to diminish unwanted autofluorescence from non-lipofuscin sources and dramatically improve signal-to-noise ratio. TrueVIEW removes unwanted autofluorescence in tissue sections due to aldehyde fixation, the presence of red blood cells, and structural elements such as collagen and elastin. The quenching action of the kit reagents provides a clear, unambiguous, “true view” localization of the target antigen.