Learn More
MSC SC Johnson Professional TruFill Hose Hook-Up Kit, ON/OFF Valve, Pressure Bleeder, Bronze Body
Catalog No. 502839800
Encompass
The TruFill Hose Hook-Up Kit is an integral component of the innovative TruFill Dispensing System, designed to simplify dilution delivery. This kit features a convenient ON/OFF valve at the dispensing end for easy control and a pressure bleeder that maintains the integrity of any vacuum breaker installed at the water source, ensuring backflow protection. It easily connects to the TruFill dispenser head via a quick-connect adapter.
  • On/off valve located at the dispensing end for controlled operation.
  • Pressure bleeder maintains integrity of vacuum breakers at water source, providing backflow protection.
  • Quick connect adapter enables easy hook-up to the dispenser head.
  • Integrates seamlessly with the dispensing system head and cartridges.
Catalog No. 50-283-9800 Supplier MSC Supplier No. 94349420
The TruFill Hose Hook-Up Kit is an integral component of the innovative TruFill Dispensing System, designed to simplify dilution delivery. This kit features a convenient ON/OFF valve at the dispensing end for easy control and a pressure bleeder that maintains the integrity of any vacuum breaker installed at the water source, ensuring backflow protection. It easily connects to the TruFill dispenser head via a quick-connect adapter.
  • On/off valve located at the dispensing end for controlled operation.
  • Pressure bleeder maintains integrity of vacuum breakers at water source, providing backflow protection.
  • Quick connect adapter enables easy hook-up to the dispenser head.
  • Integrates seamlessly with the dispensing system head and cartridges.