Learn More
Glass Expansion TruFlo Sample Monitor for HF
SDP
Catalog No. 501907284
Encompass_Preferred
TruFlo Sample Monitor 0-4.0 mL/min for HF. This TruFlo Sample Monitor provides a measurable flow range of 0-4 mL/min, a calibrated flow range is 0.2-4.0 mL/min, & has an internal volume of 0.12 mL. The sample path is made from HF resistant materials. The TruFlo is an indispensable diagnostic tool providing continuous real-time sample flow measurement, sounds out-of-range alarm, stores 24 hours of data, is ideal for sample introduction by peristaltic pump or natural aspiration, & is suitable for all ICP-OES and ICP-MS instruments. Do you ever need to repeat the analysis of a sample due to a blocked nebulizer, worn peristaltic pump tubing, or incorrect pressure of the pump tubing clamp? With the digital display of the TruFlo Sample Monitor, you always know the actual rate of sample uptake to your nebulizer. For routine aqueous and non-HF compatible models see 501907281, 501907282, and 501907283.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7284 Supplier Glass Expansion Supplier No. 708030890
TruFlo Sample Monitor 0-4.0 mL/min for HF. This TruFlo Sample Monitor provides a measurable flow range of 0-4 mL/min, a calibrated flow range is 0.2-4.0 mL/min, & has an internal volume of 0.12 mL. The sample path is made from HF resistant materials. The TruFlo is an indispensable diagnostic tool providing continuous real-time sample flow measurement, sounds out-of-range alarm, stores 24 hours of data, is ideal for sample introduction by peristaltic pump or natural aspiration, & is suitable for all ICP-OES and ICP-MS instruments. Do you ever need to repeat the analysis of a sample due to a blocked nebulizer, worn peristaltic pump tubing, or incorrect pressure of the pump tubing clamp? With the digital display of the TruFlo Sample Monitor, you always know the actual rate of sample uptake to your nebulizer. For routine aqueous and non-HF compatible models see 501907281, 501907282, and 501907283.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001