Learn More
Selleck Chemical LLC TRULI-E1061-5MG
SDP
Supplier:  Selleck Chemical LLC E10615MG
Encompass_Preferred
TRULI inhibits both Lats1 and Lats2 with a IC50 of 0 2 nM suppresses Yap phosphorylation induces cell proliferation in several cell lines and tissues and promotes the initial stages of proliferative regeneration of the sensory receptors in the inner ear
Catalog No. 50-239-0051