Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Trump's Fixative Prefills
SDP

Catalog No. 21033373
Change view
Click to view available options
Form:
Solution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form
21-033-373 Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-033-373 Supplier StatLab Supplier No. FXTRU5CS
Add to Cart
Add to Cart

A glutaraldehyde based tissue fixative commonly used in electron microscopy

  • Trumps Fixative is primarily used in histology analysis for its ability to ensure cytoplasmic organelles are thoroughly fixed and clearly defined, while other subcellular structures are visualized with high accuracy
  • This makes it an excellent choice for electron microscopy examinations
  • The fixative eco-responsible approach to the preparation of biological samples for electron microscopy ensures consistent results and is a valuable asset for laboratory needs

Specifications

For Use With (Equipment) All Tissue Processors
Grade Chemical Solution
Product Type Fixative
Solution Type Fixative
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.