Learn More
Electron Microscopy Sciences Peel Away Disposable Embedding Mold Truncated Shape 8 x 8 mm
SDP
Catalog No. 5027734
Encompass_Preferred
Peel Away Disposable Embedding Molds are used for plastic or paraffin embedding. No block trimming needed. Inside fingers hold ID slip which becomes embedded in the block for positive permanent identification.
Size: 8 x 8 mm
Catalog No. 50-277-34 Supplier Electron Microscopy Sciences Supplier No. 70180
Peel Away Disposable Embedding Molds are used for plastic or paraffin embedding. No block trimming needed. Inside fingers hold ID slip which becomes embedded in the block for positive permanent identification.
Size: 8 x 8 mm