Learn More
Alkali Scientific 200µl XL (91mm) PurePoint™ Extended Length Filter Pipet Tips, Sterile, (10 racks of 96), 960/cs
SDP
Catalog No. NC1923879
Encompass_Preferred
PurePoint™ Filter Pipet Tips are designed and manufactured using premium-grade 100% polypropylene for the most critical molecular biology liquid handling applications. All PurePoint™ Aerosol Filter Tips are gamma irradiated sterile and certified RNase/DNase-Free, human DNA-Free, and pyrogen-free. In addition, PurePoint™ Aerosol Filter Tips are Ultra-Clear with a Fine-Point design for the convenient, precise pipetting of precious nucleic acid and protein samples and are designed to be Universal-fit for the majority of research-grade pipettors*. *Not suitable for Rainin LTS (Light Touch) Pipettors.
Catalog No. NC1923879 Supplier Alkali Scientific Supplier No. FT1300
PurePoint™ Filter Pipet Tips are designed and manufactured using premium-grade 100% polypropylene for the most critical molecular biology liquid handling applications. All PurePoint™ Aerosol Filter Tips are gamma irradiated sterile and certified RNase/DNase-Free, human DNA-Free, and pyrogen-free. In addition, PurePoint™ Aerosol Filter Tips are Ultra-Clear with a Fine-Point design for the convenient, precise pipetting of precious nucleic acid and protein samples and are designed to be Universal-fit for the majority of research-grade pipettors*. *Not suitable for Rainin LTS (Light Touch) Pipettors.