Learn More
Thermo Scientific™ TruScan™ RM Next Generation Handheld Raman Analyzer
Catalog No. 17375100 Shop All Thermo Scientific Products
Change view
Click to view available options
Certifications/Compliance:
21 CFR Part 11 and GMP Compliance
Catalog No. Certifications/Compliance
17-375-100 21 CFR Part 11 and GMP Compliance
1 options
Catalog No. 17-375-100 Supplier Thermo Scientific™ Supplier No. 800003912.8
Add to Cart
Add to Cart
Request a Quote
Performs rapid material identification at the point-of-need within seconds to decrease sampling costs and increase inventory turns.
The TruScan RM analyzer offers an unparalleled solution for rapid material identification.
  • Features highly selective Raman spectroscopy
  • Fast, point of need raw material identification (10-15 seconds)
  • Portable handheld and weighs less than 2 lb. (0.9 kg)
  • Clear PASS or FAIL results enable user independence and require no secondary interpretation
  • Broad compound coverage includes cellulose based products
  • Fast and easy method development through assisted signature/reference acquisition
  • Intuitive workflow adapted to production environment through batch reports and ease of data input using barcode scanner
  • Greatly reduces analysis cost and overall overhead
  • Enhanced 21 CFR Part 11 and GMP compliance with biometric login, complex password options, and full audit trail features
  • Non-contact analysis through plastic bags, glass containers, blister packs, and clear gel caps greatly eliminates potential staff exposure to hazardous chemicals and highly potent active pharmaceutical ingredients (API) during inspection
  • Easy fleet management feature enable instrument and identification methods cloning
  • Battery operated instrument (4 hours on a single charge)
  • Qualification documentation and standard operating procedures available
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance 21 CFR Part 11 and GMP Compliance
Weight (English) < 2 lb.
Weight (Metric) < 0.9 kg