Learn More
MSC SC Johnson Professional Hospital Disinfectant, 10 oz., Concentrate, 4/Case
Catalog No. 502818223
Encompass
This one-step virucide and disinfectant effectively cleans and disinfects, offering broad-spectrum efficacy against bacteria, viruses, and fungi, while inhibiting mold and mildew growth. It is included on EPA's List Q for disinfectants against Monkeypox.
  • One-step spray and wipe virucide and disinfectant
  • Broad-spectrum efficacy against bacteria, viruses, and fungi
  • Inhibits mold and mildew growth
  • On EPA's List Q for disinfectants against Monkeypox with 5-minute contact time
  • No mixing or measuring required
  • Portable, no-spill cartridge with integrated metering system
  • Each cartridge yields up to 15 in-use quarts
  • Designed for use with mobile dispensing systems
Catalog No. 50-281-8223 Supplier MSC Supplier No. 18261750
This one-step virucide and disinfectant effectively cleans and disinfects, offering broad-spectrum efficacy against bacteria, viruses, and fungi, while inhibiting mold and mildew growth. It is included on EPA's List Q for disinfectants against Monkeypox.
  • One-step spray and wipe virucide and disinfectant
  • Broad-spectrum efficacy against bacteria, viruses, and fungi
  • Inhibits mold and mildew growth
  • On EPA's List Q for disinfectants against Monkeypox with 5-minute contact time
  • No mixing or measuring required
  • Portable, no-spill cartridge with integrated metering system
  • Each cartridge yields up to 15 in-use quarts
  • Designed for use with mobile dispensing systems