Learn More
Waters Corp TruView LCMS Certified Clear Glass 12 x 32 mm Screw Neck Total Recovery Vial, with Cap and Preslit PTFE/Silicone Septum, 1 mL Volume, 100/pk
Catalog No. NC0224966 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
TruView LCMS Certified vials are produced following tightly controlled manufacturing processes and handling procedures. They are tested for cleanliness by mass spectrometers and for low analyte adsorption by ACQUITY/MS/MS.
Catalog No. NC0224966 Supplier Waters Corp Supplier No. 186005663CV
TruView LCMS Certified vials are produced following tightly controlled manufacturing processes and handling procedures. They are tested for cleanliness by mass spectrometers and for low analyte adsorption by ACQUITY/MS/MS.