Learn More
Waters Corp TruView Vial, pH Control LCMS Certified, Clear Glass, 12 x 32 mm, 1.5 mL
Catalog No. 50179766 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These pH Control LCMS Certified Vials are manufactured to ISO 9001 standards with tightly controlled processes, ensuring cleanliness, low analyte adsorption, and chemical inertness for reliable LC and LC/MS analysis. The Max Recovery design features wide neck openings and eliminates the need for low-volume inserts.
  • LCMS certified with pH control
  • Tested for cleanliness by mass spectrometers and low analyte adsorption by LC-MS/MS
  • Chemically tested and certified for LC and LC/MS analysis
  • Certified to be within dimensional tolerances for autosamplers
  • Screw cap ensures secure septum retention and excellent seal
  • Preslit PTFE/Silicone septa for outstanding resealing, multiple injections, and temperature range from -40°C to 200°C
Catalog No. 50-179-766 Supplier Waters Corp Supplier No. 186005662CV
These pH Control LCMS Certified Vials are manufactured to ISO 9001 standards with tightly controlled processes, ensuring cleanliness, low analyte adsorption, and chemical inertness for reliable LC and LC/MS analysis. The Max Recovery design features wide neck openings and eliminates the need for low-volume inserts.
  • LCMS certified with pH control
  • Tested for cleanliness by mass spectrometers and low analyte adsorption by LC-MS/MS
  • Chemically tested and certified for LC and LC/MS analysis
  • Certified to be within dimensional tolerances for autosamplers
  • Screw cap ensures secure septum retention and excellent seal
  • Preslit PTFE/Silicone septa for outstanding resealing, multiple injections, and temperature range from -40°C to 200°C