Learn More
Waters Corp Waters Autosampler Vial, TruView pH Control LCMS Certified, Clear Glass, 2 mL
Catalog No. 50179768 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters TruView pH Control LCMS Certified Vials are manufactured following tightly controlled processes, undergoing testing for cleanliness by mass spectrometers and low analyte adsorption by LC-MS/MS. These autosampler vials are designed to withstand high temperatures and are compatible with automated systems, providing a complete solution for sample storage and analysis.
  • TruView pH Control LCMS Certified.
  • Features a screw neck closure.
  • Measures 12 x 32 mm.
  • Offers a 2 mL volume capacity.
  • Constructed from clear glass.
  • Equipped with a preslit PTFE/silicone seal.
  • Supplied with vial, cap, and septum.
  • Compatible with automated systems and autosamplers.
  • Designed for high temperature resistance.
Catalog No. 50-179-768 Supplier Waters Corp Supplier No. 186005666CV
Waters TruView pH Control LCMS Certified Vials are manufactured following tightly controlled processes, undergoing testing for cleanliness by mass spectrometers and low analyte adsorption by LC-MS/MS. These autosampler vials are designed to withstand high temperatures and are compatible with automated systems, providing a complete solution for sample storage and analysis.
  • TruView pH Control LCMS Certified.
  • Features a screw neck closure.
  • Measures 12 x 32 mm.
  • Offers a 2 mL volume capacity.
  • Constructed from clear glass.
  • Equipped with a preslit PTFE/silicone seal.
  • Supplied with vial, cap, and septum.
  • Compatible with automated systems and autosamplers.
  • Designed for high temperature resistance.