Learn More
Waters Corp Waters Vial, TruView pH Control LCMS Certified, Clear Glass, 12 x 32 mm, 2 mL
Catalog No. 50179764 Shop All Waters Corporation Products
Encompass_Preferred
TruView pH Control LCMS Certified Vials are manufactured under tightly controlled processes and handling procedures. They undergo testing for cleanliness using mass spectrometers and for low analyte adsorption through LC-MS/MS.
  • Manufactured under tightly controlled processes and handling procedures.
  • Tested for cleanliness using mass spectrometers.
  • Tested for low analyte adsorption by LC-MS/MS.
  • Features clear glass material.
  • Measures 12 x 32 mm.
  • Offers a 2 mL volume capacity.
  • Equipped with a screw neck closure.
  • Includes vial, cap, & PTFE/silicone septum.
  • Ideal for sample preparation applications.
  • Supplied in packages of 100 units.
Catalog No. 50-179-764 Supplier Waters Corp Supplier No. 186005660CV
TruView pH Control LCMS Certified Vials are manufactured under tightly controlled processes and handling procedures. They undergo testing for cleanliness using mass spectrometers and for low analyte adsorption through LC-MS/MS.
  • Manufactured under tightly controlled processes and handling procedures.
  • Tested for cleanliness using mass spectrometers.
  • Tested for low analyte adsorption by LC-MS/MS.
  • Features clear glass material.
  • Measures 12 x 32 mm.
  • Offers a 2 mL volume capacity.
  • Equipped with a screw neck closure.
  • Includes vial, cap, & PTFE/silicone septum.
  • Ideal for sample preparation applications.
  • Supplied in packages of 100 units.