Learn More
Waters Corp Waters Autosampler Vial, TruView LCMS Certified, Amber Glass, 12 x 32 mm, 2 mL
Catalog No. 50179765 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
TruView pH Control LCMS Certified Vials undergo rigorous testing for cleanliness and low analyte adsorption. This amber glass vial protects sensitive samples from light, offering compositional stability and chemical inertness for reliable analysis.
  • Amber glass for light-sensitive materials.
  • Stable composition, resistant to chemical and mechanical forces.
  • Chemically inert, prevents interference.
  • Universal screw cap provides secure seal.
  • Preslit PTFE/Silicone septum for excellent resealing.
  • Certified for LC and LC/MS analysis.
  • Batch tested for cleanliness and quality.
Catalog No. 50-179-765 Supplier Waters Corp Supplier No. 186005661CV
TruView pH Control LCMS Certified Vials undergo rigorous testing for cleanliness and low analyte adsorption. This amber glass vial protects sensitive samples from light, offering compositional stability and chemical inertness for reliable analysis.
  • Amber glass for light-sensitive materials.
  • Stable composition, resistant to chemical and mechanical forces.
  • Chemically inert, prevents interference.
  • Universal screw cap provides secure seal.
  • Preslit PTFE/Silicone septum for excellent resealing.
  • Certified for LC and LC/MS analysis.
  • Batch tested for cleanliness and quality.