Learn More
Waters Corp TruView Autosampler Vial, pH Control LCMS Certified, Amber Glass, 1.5 mL, 12 x 32 mm
Catalog No. 502037709 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These pH Control LCMS Certified Vials are manufactured to stringent quality control standards, ensuring cleanliness and low analyte adsorption. The amber glass prevents deterioration of light-sensitive materials and provides compositional stability and chemical inertness. A universal screw cap with preslit PTFE/Silicone septa creates an excellent seal.
  • Certified for pH control LCMS.
  • Amber glass prevents deterioration of lab materials.
  • Offers compositional stability and chemical inertness.
  • Stringent quality control and handling methods.
  • Tested for cleanliness and low analyte adsorption.
  • Made to ISO 9001 standards.
  • Certified to autosampler-specific dimensional tolerances.
  • Universal screw cap creates excellent seal.
  • Preslit PTFE/Silicone septa for resealing capabilities.
  • Color-coded for easy sample tracking.
Catalog No. 50-203-7709 Supplier Waters Corp Supplier No. 186005670CV
These pH Control LCMS Certified Vials are manufactured to stringent quality control standards, ensuring cleanliness and low analyte adsorption. The amber glass prevents deterioration of light-sensitive materials and provides compositional stability and chemical inertness. A universal screw cap with preslit PTFE/Silicone septa creates an excellent seal.
  • Certified for pH control LCMS.
  • Amber glass prevents deterioration of lab materials.
  • Offers compositional stability and chemical inertness.
  • Stringent quality control and handling methods.
  • Tested for cleanliness and low analyte adsorption.
  • Made to ISO 9001 standards.
  • Certified to autosampler-specific dimensional tolerances.
  • Universal screw cap creates excellent seal.
  • Preslit PTFE/Silicone septa for resealing capabilities.
  • Color-coded for easy sample tracking.