Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Truxene 98.0+%, TCI America™
SDP

Catalog No. T2752100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2752100MG 100 mg
T27521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T2752100MG Supplier TCI America Supplier No. T2752100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 548-35-6
Molecular Formula C27H18
Molecular Weight (g/mol) 342.441
MDL Number MFCD00021180
InChI Key YGPLLMPPZRUGTJ-UHFFFAOYSA-N
PubChem CID 68355
SMILES C1C2=CC=CC=C2C3=C1C4=C(CC5=CC=CC=C54)C6=C3CC7=CC=CC=C76

Specifications

Melting Point 370°C
Color Yellow
Quantity 100 mg
Synonym truxene, truxen, .alpha.-truxene, 5h-tribenzo a,f,k trindene, 10,15-dihydro, benzene, tribenzylene, 10,15-dihydro-5h-tribenzo a,f,k triindene, 10,15-dihydro-5h-diindeno 1,2-a:1',2'-c fluorene, 5h-tribenzo a,f,k trindene,10,15-dihydro, tribenzylenebenzene
Formula Weight 342.44
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Truxene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.