Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Truxenone 98.0+%, TCI America™
SDP

Catalog No. T3249200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3249200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T3249200MG Supplier TCI America Supplier No. T3249200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4430-15-3
Molecular Formula C27H12O3
Molecular Weight (g/mol) 384.39
MDL Number MFCD07808465
InChI Key HSMDIUMGCHZHNI-UHFFFAOYSA-N
Synonym Tribenzoylenebenzene, 5H-Tribenzo[a,f,k]trindene-5,10,15-trione, 5H-Diindeno[1,2-a:1′C,2′C-c]fluorene-5,10,15-trione
PubChem CID 380507
IUPAC Name heptacyclo[18.7.0.0²,¹⁰.0³,⁸.0¹¹,¹⁹.0¹²,¹⁷.0²¹,²⁶]heptacosa-1(20),2(10),3,5,7,11(19),12,14,16,21(26),22,24-dodecaene-9,18,27-trione
SMILES O=C1C2=CC=CC=C2C2=C1C1=C(C(=O)C3=CC=CC=C13)C1=C2C(=O)C2=C1C=CC=C2

Specifications

Melting Point 367°C
Quantity 200 mg
Formula Weight 384.39
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Truxenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.