Learn More
Medchemexpress LLC HY-P2141A 1mg Medchemexpress, TRV-120027 (TFA) CAS:0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP2141A1MG
Encompass_Preferred
Medchemexpress, HY-P2141A 1mg TRV-120027 (TFA) CAS:0 TRV120027 TFA, a β-arrestin-1-biased agonist of the angiotensin II receptor type 1 (AT1R), engages ß-arrestins while blocking G-protein signaling. TRV120027 TFA induces acute catecholamine secretion through cation channel subfamily C3 (TRPC3) coupling, promotes the formation of a macromolecular complex composed of AT1R–β-arrestin-1–TRPC3–PLCγ at the plasma membrane. TRV120027 TFA inhibits angiotensin II–mediated vasoconstriction and increases cardiomyocyte contractility. TRV120027 TFA has the potential for the acute decompensated heart failure (ADHF) treatment[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1709