Learn More
Enzo Life Sciences Thioredoxin (E. coli), (recombinant) (His-tag) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPP892200
Encompass_Preferred
Alternative name: Trx. Purity: ≥95% (SDS-PAGE; Western blot). MW: ~12kDa. Formulation: Liquid. In 50mM TRIS, pH 7.5, containing 100mM sodium chloride and 1mM EDTA. Source: Produced in E. coli. E. coli thioredoxin is fused at the N-terminus to a His-tag. Long Term Storage: -80°C. Applications: WB Activity assay.
Catalog No. 50-200-7654