Learn More
Cayman Chemical TRx0237 (mesylate), Purity: ≥95%, CAS: 1236208-20-0, 10 mg
SDP
Catalog No. 502946741
Encompass_Preferred
An inhibitor of tau aggregation; inhibits tau-tau binding (IC50 = 238 µM) and tau aggregation in 3T6H cells expressing human tau (EC50 = 0.19 µM); increases hippocampal ACh and synaptophysin levels, as well as increases mitochondrial complex IV activity, decreases tau pathology, and reverses deficits in spatial learning in the L1 transgenic mouse model of Alzheimer’s disease
Catalog No. 50-294-6741 Supplier Cayman Chemical Supplier No. 3430710 MG
An inhibitor of tau aggregation; inhibits tau-tau binding (IC50 = 238 µM) and tau aggregation in 3T6H cells expressing human tau (EC50 = 0.19 µM); increases hippocampal ACh and synaptophysin levels, as well as increases mitochondrial complex IV activity, decreases tau pathology, and reverses deficits in spatial learning in the L1 transgenic mouse model of Alzheimer’s disease