Learn More
Cayman Chemical TRx0237 (mesylate), Purity: ≥95%, CAS: 1236208-20-0, 50 mg
SDP
Catalog No. 502946821
Encompass_Preferred
An inhibitor of tau aggregation; inhibits tau-tau binding (IC50 = 238 µM) and tau aggregation in 3T6H cells expressing human tau (EC50 = 0.19 µM); increases hippocampal ACh and synaptophysin levels, as well as increases mitochondrial complex IV activity, decreases tau pathology, and reverses deficits in spatial learning in the L1 transgenic mouse model of Alzheimer’s disease
Catalog No. 50-294-6821 Supplier Cayman Chemical Supplier No. 3430750 MG
An inhibitor of tau aggregation; inhibits tau-tau binding (IC50 = 238 µM) and tau aggregation in 3T6H cells expressing human tau (EC50 = 0.19 µM); increases hippocampal ACh and synaptophysin levels, as well as increases mitochondrial complex IV activity, decreases tau pathology, and reverses deficits in spatial learning in the L1 transgenic mouse model of Alzheimer’s disease