Learn More
Cayman Chemical TrxR Assay Buffer (10X), 1 ea
SDP
Catalog No. 502887088
Encompass_Preferred
TrxR Assay Buffer (10X) has been tested and formulated to work exclusively with Cayman’s Thioredoxin Reductase Colorimetric Assay Kit (Item No. 10007892).
Catalog No. 50-288-7088 Supplier Cayman Chemical Supplier No. 100090921 EA
TrxR Assay Buffer (10X) has been tested and formulated to work exclusively with Cayman’s Thioredoxin Reductase Colorimetric Assay Kit (Item No. 10007892).