Learn More
Cayman Chemical TrxR NADPH, 1 ea
SDP
Catalog No. 502886931
Encompass_Preferred
TrxR NADPH has been tested and formulated to work exclusively with Cayman’s Thioredoxin Reductase Colorimetric Assay Kit (Item No. 10007892).
Catalog No. 50-288-6931 Supplier Cayman Chemical Supplier No. 100090951 EA
TrxR NADPH has been tested and formulated to work exclusively with Cayman’s Thioredoxin Reductase Colorimetric Assay Kit (Item No. 10007892).