Learn More
NEOGEN Tryptic Soy Agar with Lecithin and Tween 80, 5 kg
SDP
Catalog No. 502015184
Encompass_Preferred
Tryptic Soy Agar with Lecithin and Tween 80 is used for the isolation of microorganisms from surfaces sanitized with quaternary ammonium compounds and is not intended for use in the diagnosis of disease or other conditions in humans. In 1955, Leavitt et al. discovered Tryptic Soy Agar supported excellent growth of aerobic and anaerobic microorganisms. Tryptic Soy Agar is a nutritious base and a variety of supplements are added to enhance the medium, including Lecithin and Tween 80. The Lecithin and Tween 80 inactivate some preservatives that may inhibit bacterial growth, reducing “preservative carryover”. Tryptic Soy Agar with Lecithin and Tween 80 is recommended for determining the sanitation efficiency of containers, equipment, and work area (environmental monitoring).For more information visit https://www.neogen.com/solutions/Microbiology/tryptic-soy-agar-with-lecithin-and-tween-80
Catalog No. 50-201-5184 Supplier NEOGEN Supplier No. 700002992
Tryptic Soy Agar with Lecithin and Tween 80 is used for the isolation of microorganisms from surfaces sanitized with quaternary ammonium compounds and is not intended for use in the diagnosis of disease or other conditions in humans. In 1955, Leavitt et al. discovered Tryptic Soy Agar supported excellent growth of aerobic and anaerobic microorganisms. Tryptic Soy Agar is a nutritious base and a variety of supplements are added to enhance the medium, including Lecithin and Tween 80. The Lecithin and Tween 80 inactivate some preservatives that may inhibit bacterial growth, reducing “preservative carryover”. Tryptic Soy Agar with Lecithin and Tween 80 is recommended for determining the sanitation efficiency of containers, equipment, and work area (environmental monitoring).For more information visit https://www.neogen.com/solutions/Microbiology/tryptic-soy-agar-with-lecithin-and-tween-80