Learn More
MilliporeSigma TRYP SOY AGAR CASSETTE 48
Catalog No. NC9412439
Encompass
Pre-filled Milliflex Cassettes, product name: Tryptic Soy Agar, target microorganisms, Aerobic Heterotrophic microorganisms, for use with the Milliflex system, 48/PK.
Catalog No. NC9412439 Supplier MilliporeSigma Supplier No. MXSMCTS48
Pre-filled Milliflex Cassettes, product name: Tryptic Soy Agar, target microorganisms, Aerobic Heterotrophic microorganisms, for use with the Milliflex system, 48/PK.