Learn More
Alkali Scientific Trypan Blue, 100 G
SDP
Catalog No. 5000661715
Encompass_Preferred
Trypan Blue, 100g is a vital dye commonly used in cell viability assays, particularly for staining dead cells. The dye is able to penetrate the membranes of dead cells, but live cells with intact membranes exclude the dye, making it a useful marker in biological studies to distinguish between living and non-living cells. It is frequently used in cell culture experiments to assess cell mortality after treatment with toxic substances. The 100g pack size is suitable for laboratories conducting large-scale studies or those requiring a substantial quantity of dye for repeated testing. This reagent is critical in cell viability research, particularly in evaluating cytotoxicity and drug testing.
Catalog No. 50-006-61715 Supplier Alkali Scientific Supplier No. RYP003CP
Trypan Blue, 100g is a vital dye commonly used in cell viability assays, particularly for staining dead cells. The dye is able to penetrate the membranes of dead cells, but live cells with intact membranes exclude the dye, making it a useful marker in biological studies to distinguish between living and non-living cells. It is frequently used in cell culture experiments to assess cell mortality after treatment with toxic substances. The 100g pack size is suitable for laboratories conducting large-scale studies or those requiring a substantial quantity of dye for repeated testing. This reagent is critical in cell viability research, particularly in evaluating cytotoxicity and drug testing.