Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trypan Blue 40% MP Biomedicals

Catalog No. ICN19553205 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19553205 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19553205 Supplier MP Biomedicals, Inc Supplier No. 0219553205
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 72-57-1
Molecular Formula C34H24N6Na4O14S4
Molecular Weight (g/mol) 960.79
MDL Number MFCD00003969
InChI Key WMYVHJWZUUEZNE-ARWFNKCKSA-J
Synonym Direct Blue 14, Niagara Blue 3B
PubChem CID 9562061
IUPAC Name tetrasodium (3Z)-5-amino-3-[2-(4'-{2-[(2Z)-8-amino-1-oxo-3,6-disulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}-3,3'-dimethyl-[1,1'-biphenyl]-4-yl)hydrazin-1-ylidene]-4-oxo-3,4-dihydronaphthalene-2,7-disulfonate
SMILES [Na+].[Na+].[Na+].[Na+].CC1=CC(=CC=C1N\N=C1\C(=O)C2=C(N)C=C(C=C2C=C1S([O-])(=O)=O)S([O-])(=O)=O)C1=CC=C(N\N=C2\C(=O)C3=C(N)C=C(C=C3C=C2S([O-])(=O)=O)S([O-])(=O)=O)C(C)=C1

Specifications

Assay Percent Range 40%
Formula Weight 960.8
Quantity 5 g
Chemical Name or Material Trypan Blue
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.