Learn More
Alkali Scientific Trypan Blue, 5 G
SDP
Catalog No. 5000661783
Encompass_Preferred
Trypan Blue is a dye used primarily in cell viability testing. This 5g container is ideal for small-scale applications where staining of cells is required to distinguish between live and dead cells. The dye is excluded from viable cells but penetrates dead cells, allowing for clear visualization of non-viable cells. It is commonly used in cytotoxicity assays, cell culture, and clinical studies to evaluate the effects of treatments on cell viability. Trypan Blue is often used in laboratories performing cell-based assays, drug testing, and research that requires accurate live/dead cell differentiation.
Catalog No. 50-006-61783 Supplier Alkali Scientific Supplier No. RYP001CP
Trypan Blue is a dye used primarily in cell viability testing. This 5g container is ideal for small-scale applications where staining of cells is required to distinguish between live and dead cells. The dye is excluded from viable cells but penetrates dead cells, allowing for clear visualization of non-viable cells. It is commonly used in cytotoxicity assays, cell culture, and clinical studies to evaluate the effects of treatments on cell viability. Trypan Blue is often used in laboratories performing cell-based assays, drug testing, and research that requires accurate live/dead cell differentiation.