Learn More
Sigma Aldrich Fine Chemicals Biosciences Trypan Blue solution 0.4%, liquid, sterile-filtered, suitable for cell culture | 72-57-1 | MFCD00003969 | 100ML
Catalog No. NC9929604
Encompass
Trypan Blue solution 0.4%, liquid, sterile-filtered, suitable for cell culture | Mol Wt: 960.81 | 72-57-1 | MFCD00003969 | 100ML
Catalog No. NC9929604 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T8154100ML
Trypan Blue solution 0.4%, liquid, sterile-filtered, suitable for cell culture | Mol Wt: 960.81 | 72-57-1 | MFCD00003969 | 100ML