Learn More
Description
Trypan Blue (C. l. 23850) is a large, tetrasulfonated anionic, hydrophilic dye. It is also known as diamine blue 3B.
Specifications
Specifications
| Chemical Name or Material | Trypan Blue |
| CAS | 72-57-1 |
| Color | Black/Brown or Black/Green to Brown |
| Quantity | 100 g |
| Molecular Weight (g/mol) | 960.81 g/mol |
| Percent Purity | ≥80% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.