Learn More
Aat Bioquest Trypan Red Plus™ *0.1 M aqueous solution*
SDP
Supplier:  Aat Bioquest 2457
Encompass_Preferred
Trypan Red Plus™ is similar to Trypan Blue in cell permeability. It is not permeable to live cells.
Catalog No. 50-195-3174