Learn More
Roche Diagnostics TRYPANBLUE SOL REFILL 4 X 60ML
Supplier:  Roche Diagnostics 05650640001
Encompass
Trypan Blue Solution is required as an exclusion stain for viability determination with the Cedex and Cedex HiRes Systems. The ready-to-use solution is available as a component of the Cedex Reagent Kit, Cedex HiRes Reagent Kit, &separately.
Catalog No. 50-589-351