Learn More
ATCC Quantitative Genomic DNA from; Leishmania major
Catalog No. 502381159
Encompass
Quantitative genomic DNA from Leishmania major can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine protozoan load.
Catalog No. 50-238-1159 Supplier ATCC Supplier No. 30012DQ
Quantitative genomic DNA from Leishmania major can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine protozoan load.